3,3-Bis(bromomethyl)oxetane 95%

Supplier: Ambeed
Warning

Synonyms: 2,2-Bis(bromomethyl)-1,3-epoxypropane

A115785-100G A115785-10G A115785-25G A115785-1G A115785-5G
103798-620EA 9 USD
103798-620 103824-480 77271-186 77271-188 77271-190 77297-394 77297-396
3,3-Bis(bromomethyl)oxetane 95%
3,3-Bis(bromomethyl)oxetane

Precaución: for Research Use Only

Formula: C₅H₈Br₂O
MW: 243.93 g/mol
Boiling Pt: 249.4 °C (760 mm Hg)
Flash Pt: 98 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00269694
CAS Number: 2402-83-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR