3,3',5,5'-Tetra-tert-butyl-4,4'-diphenoquinone 98%

Supplier: Ambeed
Warning

Synonyms: 3,3',5,5'-Tetra-tert-butyl-[1,1'-bi(cyclohexylidene)]-2,2',5,5'-tetraene-4,4'-dione, 3,3',5,5'-Tetra-tert-butyldiphenoquinone

A124761-100G A124761-10G A124761-1G A124761-25G A124761-5G
76996-042EA 170.81 USD
76996-042 76996-044 76996-046 76996-048 77312-716
3,3',5,5'-Tetra-tert-butyl-4,4'-diphenoquinone 98%
3,3',5,5'-Tetra-tert-butyl-4,4'-diphenoquinone

Precaución: for Research Use Only

Formula: C₂₈H₄₀O₂
MW: 408.63 g/mol
Boiling Pt: 492.6 °C (760 mm Hg)
Melting Pt: 242…246 °C
Flash Pt: 182.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00051798
CAS Number: 2455-14-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR