3,3',5,5'-Tetramethyl-1H,1'H-4,4'-bipyrazole 98%

Supplier: Ambeed
Warning

A121523-100MG A121523-250MG A121523-5G A121523-1G
103800-084EA 90 USD
103800-084 76822-444 76822-446 76822-448 77312-204
3,3',5,5'-Tetramethyl-1H,1'H-4,4'-bipyrazole 98%
3,3',5,5'-Tetramethyl-1H,1'H-4,4'-bipyrazole

Precaución: for Research Use Only

Formula: C₁₀H₁₄N₄
MW: 190.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00464842
CAS Number: 4054-67-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR