4,4'-Dihydroxy-3,3',5,5'-tetraisopropylbiphenyl 97%
Supplier: Ambeed
|
Warning
|
A185075-250MG
A185075-1G
A185075-100MG
A185075-10G
103815-818EA
140
USD
103815-818
103846-384
76813-714
77301-946
77881-392
77881-393
4,4'-Dihydroxy-3,3',5,5'-tetraisopropylbiphenyl 97%
4,4'-Dihydroxy-3,3',5,5'-tetraisopropylbiphenyl
Precaución: for Research Use Only
Formula:
C₂₄H₃₄O₂ Storage Temperature: Ambient |
MDL Number:
MFCD20257818 CAS Number: 2416-95-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...