1H,1H,2H,2H-Tridecafluoro-n-octyl acrylate 95% stabilized

Supplier: Ambeed
Warning

Synonyms: 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluorooctyl acrylate, Acrylic acid 1H,1H,2H,2H-Tridecafluoro-n-octyl ester

Stabilization: Stabilized with MEHQ or TBC

A137864-100G A137864-10G A137864-500G A137864-25G A137864-5G
103802-522EA 10 USD
103802-522 77234-772 77234-774 77234-776 77315-508 77315-510
1H,1H,2H,2H-Tridecafluoro-n-octyl acrylate 95% stabilized
1H,1H,2H,2H-Tridecafluoro-n-octyl acrylate

Precaución: for Research Use Only

Formula: C₁₁H₇F₁₃O₂
MW: 418.154 g/mol
Flash Pt: 102 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00042351
CAS Number: 17527-29-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR