3,3,3-Trifluoropropan-1-amine hydrochloride 98%

Supplier: Ambeed
Warning

A255145-100MG A255145-10G A255145-250MG A255145-1G A255145-5G A255145-25G
103812-184EA 37 USD
103812-184 103852-038 76976-778 76976-780 76976-782 77091-498 77091-500 77612-446
3,3,3-Trifluoropropan-1-amine hydrochloride 98%
3,3,3-Trifluoropropan-1-amine hydrochloride

Precaución: for Research Use Only

Formula: C3H7ClF3N
Melting Pt: 222…223 °C
Storage Temperature: Ambient
MDL Number: MFCD00236725
CAS Number: 2968-33-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR