3,3,3-Trifluoro-2-methylpropanoic acid 97%

Supplier: Ambeed
Warning

A172164-100MG A172164-250MG A172164-1G A172164-5G
103806-704EA 195 USD
103806-704 103844-988 76824-546 76824-548 77082-172 77082-174
3,3,3-Trifluoro-2-methylpropanoic acid 97%
3,3,3-Trifluoro-2-methylpropanoic acid

Precaución: for Research Use Only

Formula: C₄H₅F₃O₂
MW: 142.077 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD19228247
CAS Number: 381-97-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR