3,3,3-Trifluoro-2-methylpropan-1-ol 97%
Supplier: Ambeed
|
Danger
|
A168581-100MG
A168581-250MG
A168581-1G
A168581-5G
103806-300EA
198
USD
103806-300
103844-554
76813-122
76813-124
77300-932
77300-934
3,3,3-Trifluoro-2-methylpropan-1-ol 97%
3,3,3-Trifluoro-2-methylpropan-1-ol
Precaución: for Research Use Only
Formula:
C₄H₇F₃O Storage Temperature: Refrigerator |
MDL Number:
MFCD20483912 CAS Number: 431-23-2 UN: 1993 ADR: 3,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...