3,3,3-Trifluoro-2,2-dimethylpropionic acid 97%

Supplier: Ambeed
Danger

Synonyms: 3,3,3-Trifluoro-2,2-dimethylpropanoic acid

A373765-1G A373765-25G A373765-5G A373765-250MG
103826-542EA 12 USD
103826-542 103837-560 76840-828 76840-830 76840-832 77278-464 77278-466
3,3,3-Trifluoro-2,2-dimethylpropionic acid 97%
3,3,3-Trifluoro-2,2-dimethylpropionic acid

Precaución: for Research Use Only

Formula: C₅H₇F₃O₂
Melting Pt: 66…71 °C
Flash Pt: 46.6 °C
Storage Temperature: Ambient
MDL Number: MFCD08445819
CAS Number: 889940-13-0
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR