3,3'-(Oxybis(methylene))bis(3-ethyloxetane) 95%

Supplier: Ambeed
Warning

A228012-100G A228012-10G A228012-25G A228012-5G A228012-1G
103819-354EA 5 USD
103819-354 103850-262 76826-382 76826-384 77089-050 77089-052 77303-812
3,3'-(Oxybis(methylene))bis(3-ethyloxetane) 95%
3,3'-(Oxybis(methylene))bis(3-ethyloxetane)

Precaución: for Research Use Only

Formula: C₁₂H₂₂O₃
MW: 214.3 g/mol
Boiling Pt: 275 °C (760 mm Hg)
Flash Pt: 91 °C
Storage Temperature: Refrigerator
MDL Number: MFCD17214770
CAS Number: 18934-00-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR