3-(Tritylthio)propanoic acid 98%

Supplier: Ambeed
Warning

Synonyms: S-Trityl-3-mercaptopropionic acid, 3-Tritylsulphanyl-propionic acid, 3-(Tritylthio)propionic acid, Mpa(Trt)

A120429-100G A120429-10G A120429-25G A120429-500G A120429-1G A120429-5G
103799-930EA 5 USD
103799-930 76995-782 76995-784 76995-786 76995-788 77297-760 77297-762
3-(Tritylthio)propanoic acid 98%
3-(Tritylthio)propanoic acid

Precaución: for Research Use Only

Formula: C₂₂H₂₀O₂S
Boiling Pt: 503.8 °C (760 mm Hg)
Flash Pt: 258.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00237291
CAS Number: 27144-18-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR