3-(Trifluoromethyl)thiophene-2-carboxylic acid 98%

Supplier: Ambeed
Warning

A167185-100MG A167185-250MG
103806-144EA 117 USD
103806-144 103844-406 76985-266 76985-268
3-(Trifluoromethyl)thiophene-2-carboxylic acid 98%
3-(Trifluoromethyl)thiophene-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₃F₃O₂S
MW: 196.15 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20657880
CAS Number: 767337-59-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR