3-(Trifluoromethyl)pyridine 98%
Supplier: Ambeed
|
Danger
|
A270544-1G
A270544-25G
A270544-5G
103813-004EA
10
USD
103813-004
103852-908
77305-056
77305-058
77305-060
3-(Trifluoromethyl)pyridine 98%
3-(Trifluoromethyl)pyridine
Precaución: for Research Use Only
Formula:
C₆H₄F₃N MW: 147.1 g/mol Boiling Pt: 127.8 °C (760 mm Hg) Flash Pt: 23 °C Storage Temperature: Ambient |
MDL Number:
MFCD00100032 CAS Number: 3796-23-4 UN: 1992 ADR: 3,6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...