3-(Trifluoromethyl)pyrazin-2-amine 95%

Supplier: Ambeed
Warning

A183745-1G A183745-5G
103815-664EA 248 USD
103815-664 103846-226 77301-850 77301-852
3-(Trifluoromethyl)pyrazin-2-amine 95%
3-(Trifluoromethyl)pyrazin-2-amine

Precaución: for Research Use Only

Formula: C₅H₄F₃N₃
MW: 163.1 g/mol
Boiling Pt: 220.8±35.0 °C (760 mm Hg)
Flash Pt: 87.3 °C
Storage Temperature: Ambient
MDL Number: MFCD15144417
CAS Number: 213019-67-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR