(α,α,α-Trifluoro-3-tolyl)acetic acid 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
(3-(Trifluoromethyl)phenyl)acetic acid, 3-(Trifluoromethyl)phenylacetic acid
A301216-100G
A301216-10G
A301216-500G
A301216-25G
A301216-5G
103835-448EA
10
USD
103835-448
76693-414
76693-416
76693-418
76838-128
77305-824
(α,α,α-Trifluoro-3-tolyl)acetic acid 95%
(α,α,α-Trifluoro-3-tolyl)acetic acid
Precaución: for Research Use Only
Formula:
(C₆H₄(CF₃))CH₂COOH MW: 204.15 g/mol Boiling Pt: 251.8 °C (760 mm Hg) Melting Pt: 76…79 °C Flash Pt: 106.1 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004339 CAS Number: 351-35-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...