α,α,α-Trifluoro-m-cresol 98%

Supplier: Ambeed
Warning

A200287-100G A200287-10G A200287-25G
103809-342EA 85 USD
103809-342 76825-548 76825-550 77302-672
α,α,α-Trifluoro-m-cresol 98%
α,α,α-Trifluoro-m-cresol

Precaución: for Research Use Only

Formula: C₇H₅F₃O
Boiling Pt: 178…179 °C (760 mm Hg)
Melting Pt: 2…1.8 °C
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00002299
CAS Number: 98-17-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR