3-(Trifluoromethyl)-1H-pyrazolo[3,4-b]pyridine 97%

Supplier: Ambeed
Warning

A132701-250MG A132701-1G
103801-762EA 18 USD
103801-762 77027-136 77314-352
3-(Trifluoromethyl)-1H-pyrazolo[3,4-b]pyridine 97%
3-(Trifluoromethyl)-1H-pyrazolo[3,4-b]pyridine

Precaución: for Research Use Only

Formula: C₇H₄F₃N₃
MW: 187.122 g/mol
Boiling Pt: 284.586 °C (760 mm Hg)
Flash Pt: 125.914 °C
Storage Temperature: Refrigerator
MDL Number: MFCD13178059
CAS Number: 956010-87-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR