3-(Trifluoromethoxy)phenol 98%

Supplier: Ambeed
Danger

Synonyms: α,α,α-Trifluoro-3-hydroxyanisole

A258779-10G A258779-1G A258779-5G
103812-398EA 9 USD
103812-398 103852-276 76874-798 77091-810 77304-750
3-(Trifluoromethoxy)phenol 98%
3-(Trifluoromethoxy)phenol

Precaución: for Research Use Only

Formula: C₇H₅F₃O₂
MW: 178.11 g/mol
Boiling Pt: 69…70 °C (12 mm Hg)
Flash Pt: 84 °C
Storage Temperature: Ambient
MDL Number: MFCD00040987
CAS Number: 827-99-6
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR