1-Bromo-3-(trifluoromethoxy)benzene 98%
Supplier: Ambeed
|
Warning
|
A335293-5G
A335293-10G
A335293-500G
103836-658EA
5
USD
103836-658
76839-672
77106-556
77106-558
1-Bromo-3-(trifluoromethoxy)benzene 98%
1-Bromo-3-(trifluoromethoxy)benzene
Precaución: for Research Use Only
Formula:
C₇H₄BrF₃O Flash Pt: 63 °C Storage Temperature: Ambient |
MDL Number:
MFCD00040943 CAS Number: 2252-44-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...