3-(Trifluoromethoxy)benzoic acid 97%

Supplier: Ambeed
Warning

A216883-100G A216883-10G A216883-1G A216883-500G A216883-25G A216883-5G
103849-310EA 15 USD
103849-310 76770-710 76770-712 76770-714 76770-716 77087-914 77087-916
3-(Trifluoromethoxy)benzoic acid 97%
3-(Trifluoromethoxy)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₅F₃O₃
MW: 206.12 g/mol
Melting Pt: 89…92 °C
Flash Pt: 101.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00041501
CAS Number: 1014-81-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR