3-(2-Thienyl)-1-propanol 95%

Supplier: Ambeed
Warning

Synonyms: 3-(Thiophen-2-yl)propan-1-ol

A373521-100MG A373521-250MG A373521-1G A373521-5G
103826-536EA 66 USD
103826-536 103837-554 77003-542 77003-544 77306-854 77306-856
3-(2-Thienyl)-1-propanol 95%
3-(2-Thienyl)-1-propanol

Precaución: for Research Use Only

Formula: C₇H₁₀OS
Boiling Pt: 240.4 °C (760 mm Hg)
Flash Pt: 99.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09927267
CAS Number: 19498-72-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR