3-tert-Butylisoxazole-5-amine 95%

Supplier: Ambeed
Warning

Synonyms: 3-(tert-Butyl)isoxazol-5-amine

A404279-1G A404279-5G A404279-100MG A404279-250MG
103827-106EA 27 USD
103827-106 103838-146 76841-626 76841-628 77291-202 77291-204
3-tert-Butylisoxazole-5-amine 95%
3-tert-Butylisoxazole-5-amine

Precaución: for Research Use Only

Formula: C₇H₁₂N₂O
MW: 140.19 g/mol
Boiling Pt: 248.1 °C (760 mm Hg)
Melting Pt: 110…114 °C
Flash Pt: 103.9 °C
Storage Temperature: Ambient
MDL Number: MFCD05266697
CAS Number: 59669-59-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR