3-(tert-Butyl)-1-(p-tolyl)-1H-pyrazol-5-amine 97%
Supplier: Ambeed
|
Warning
|
A429531-1G
A429531-25G
A429531-5G
A429531-250MG
103827-504EA
82
USD
103827-504
103838-560
76842-244
76842-246
76842-248
77168-612
3-(tert-Butyl)-1-(p-tolyl)-1H-pyrazol-5-amine 97%
3-(tert-Butyl)-1-(p-tolyl)-1H-pyrazol-5-amine
Precaución: for Research Use Only
Formula:
C14H19N3 MW: 229.3208 g/mol Boiling Pt: 368.3 °C (760 mm Hg) Storage Temperature: Ambient |
MDL Number:
MFCD04115090 CAS Number: 285984-25-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...