3-(tert-Butoxycarbonylamino)-3-methylbutanoic acid 97%
Supplier: Ambeed
|
Warning
|
A364700-100MG
A364700-10G
A364700-250MG
A364700-5G
103826-368EA
24
USD
103826-368
103837-388
77290-804
77290-806
77290-808
77306-762
3-(tert-Butoxycarbonylamino)-3-methylbutanoic acid 97%
3-(tert-Butoxycarbonylamino)-3-methylbutanoic acid
Precaución: for Research Use Only
Formula:
C10H19NO4 MW: 217.2600 g/mol Boiling Pt: 346.5±25.0 °C (760 mm Hg) Flash Pt: 163.3 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD09031500 CAS Number: 129765-95-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...