3-(2-Pyrrolidinyl)pyridine 97%

Supplier: Ambeed
Warning

Synonyms: 2-(3-Pyridyl)pyrrolidine, (±)-Nornicotine, DL-Nornicotine, 3-(Pyrrolidin-2-yl)pyridine

A348497-10G A348497-1G A348497-5G A348497-250MG
103825-992EA 43 USD
103825-992 103836-982 77251-498 77290-652 77290-654 77616-524
3-(2-Pyrrolidinyl)pyridine 97%
3-(2-Pyrrolidinyl)pyridine

Precaución: for Research Use Only

Formula: C₉H₁₂N₂
MW: 148.21 g/mol
Boiling Pt: 107…108 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00016913
CAS Number: 5746-86-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR