3-Pyrid-4-ylbenzoic acid 95%

Supplier: Ambeed
Warning

Synonyms: 3-(Pyridin-4-yl)benzoic acid

A239991-250MG A239991-1G A239991-10G
103811-326EA 48 USD
103811-326 103851-124 77288-710 77304-182 77748-422
3-Pyrid-4-ylbenzoic acid 95%
3-Pyrid-4-ylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₂H₉NO₂
MW: 199.2054 g/mol
Boiling Pt: 405 °C (760 mm Hg)
Melting Pt: 268 °C
Flash Pt: 198.7 °C
Storage Temperature: Ambient
MDL Number: MFCD05864808
CAS Number: 4385-78-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR