3-(3-Pyridyl)propiolic acid 95%

Supplier: Ambeed
Warning

Synonyms: 3-(Pyridin-3-yl)propiolic acid

A625596-100MG A625596-250MG A625596-1G A625596-5G
103840-244EA 1198 USD
103840-244 76819-564 76819-566 77307-994 77307-996
3-(3-Pyridyl)propiolic acid 95%
3-(3-Pyridyl)propiolic acid

Precaución: for Research Use Only

Formula: C₈H₅NO₂
MW: 147.131 g/mol
Boiling Pt: 341.4 °C (760 mm Hg)
Flash Pt: 160.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02177466
CAS Number: 59608-01-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR