3-(Pyridin-3-yl)aniline 95%

Supplier: Ambeed
Warning

Synonyms: 3-(Pyridin-3-yl)benzenamine

A245015-1G A245015-100MG A245015-250MG A245015-5G
103811-618EA 59 USD
103811-618 103851-452 77090-682 77288-832 77288-834 77304-346
3-(Pyridin-3-yl)aniline 95%
3-(Pyridin-3-yl)aniline

Precaución: for Research Use Only

Formula: C₁₁H₁₀N₂
MW: 170.211 g/mol
Boiling Pt: 368.54 °C (760 mm Hg)
Melting Pt: 74…78 °C
Flash Pt: 204.357 °C
Storage Temperature: Ambient
MDL Number: MFCD07368549
CAS Number: 57976-57-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR