3-Pyrid-2-ylbenzoic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-(Pyridin-2-yl)benzoic acid

A153785-1G A153785-5G A153785-250MG
103804-568EA 39 USD
103804-568 103842-746 77318-426 77318-428 77611-456
3-Pyrid-2-ylbenzoic acid 98%
3-Pyrid-2-ylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₂H₉NO₂
MW: 199.2054 g/mol
Boiling Pt: 409 °C (760 mm Hg)
Melting Pt: 207 °C
Flash Pt: 201.1 °C
Storage Temperature: Ambient
MDL Number: MFCD05864807
CAS Number: 4467-07-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR