1-(3-sulfopropyl)pyridinium hydroxide inner salt 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
NDSB 201, 3-(1-Pyridinio)propanesulphonate, 3-(Pyridin-1-ium-1-yl)propane-1-sulphonate
A109116-100G
A109116-25G
A109116-500G
103808-012EA
169
USD
103808-012
103823-236
76995-096
76995-098
76995-100
1-(3-sulfopropyl)pyridinium hydroxide inner salt 97%
1-(3-sulfopropyl)pyridinium hydroxide inner salt
Precaución: for Research Use Only
Formula:
C₈H₁₁NO₃S MW: 201.24 g/mol Melting Pt: 280 °C Flash Pt: 160 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064468 CAS Number: 15471-17-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...