3-Piperidin-4-ylpropan-1-ol 97%

Supplier: Ambeed
Warning

Synonyms: 3-(Piperidin-4-yl)propan-1-ol

A284728-100G A284728-250MG A284728-1G
103813-648EA 19 USD
103813-648 103834-780 76723-374 76723-376 76837-306
3-Piperidin-4-ylpropan-1-ol 97%
3-Piperidin-4-ylpropan-1-ol

Precaución: for Research Use Only

Formula: C₈H₁₇NO
MW: 143.23 g/mol
Boiling Pt: 235.5 °C (760 mm Hg)
Flash Pt: 90 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02178380
CAS Number: 7037-49-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR