3-(Piperidin-4-yl)indolin-2-one hydrochloride 95%

Supplier: Ambeed
Warning

A447940-1G A447940-100MG
103827-770EA 177 USD
103827-770 76842-644 76990-688
3-(Piperidin-4-yl)indolin-2-one hydrochloride 95%
3-(Piperidin-4-yl)indolin-2-one hydrochloride

Precaución: for Research Use Only

Formula: C₁₃H₁₇ClN₂O
MW: 252.73 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16547733
CAS Number: 79098-85-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR