3-(Phenylsulfonyl)propanoic acid 97%

Supplier: Ambeed
Warning

A392541-10G A392541-1G
103837-938EA 25 USD
103837-938 77278-646 77278-648
3-(Phenylsulfonyl)propanoic acid 97%
3-(Phenylsulfonyl)propanoic acid

Precaución: for Research Use Only

Formula: C₉H₁₀O₄S
Boiling Pt: 458.9 °C (760 mm Hg)
Melting Pt: 128…130 °C (760 mm Hg)
Flash Pt: 231.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00010141
CAS Number: 10154-71-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR