3-Phenylamino-propionic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-(Phenylamino)propanoic acid

A136384-1G A136384-5G
103802-296EA 16 USD
103802-296 77298-846 77298-848
3-Phenylamino-propionic acid 97%
3-Phenylamino-propionic acid

Precaución: for Research Use Only

Formula: C₉H₁₁NO₂
MW: 165.189 g/mol
Boiling Pt: 364.6 °C (760 mm Hg)
Melting Pt: 218…221 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00089905
CAS Number: 5652-38-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR