3-(N-tert-Butoxycarbonyl-N-methylamino)aniline 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-(N-Tert-butoxycarbonyl-N-methylamino)aniline, N-Methylbenzene-1,3-diamine, N-BOC protected
A122650-100MG
A122650-10G
A122650-1G
A122650-5G
103800-246EA
106
USD
103800-246
76971-640
76971-642
77297-878
77297-880
3-(N-tert-Butoxycarbonyl-N-methylamino)aniline 97%
3-(N-tert-Butoxycarbonyl-N-methylamino)aniline
Precaución: for Research Use Only
Formula:
C12H18N2O2 MW: 222.28 g/mol Boiling Pt: 347.8 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD13677178 CAS Number: 528882-16-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...