Sodium-3-[ethyl(m-tolyl)amino]-2-hydroxy-1-propanesulfonate hydrate 98%

Supplier: Ambeed
Warning

A366942-100G A366942-100MG A366942-5G A366942-1G A366942-25G
103826-416EA 11 USD
103826-416 76788-624 76788-626 76788-628 77306-788 77306-790
Sodium-3-[ethyl(m-tolyl)amino]-2-hydroxy-1-propanesulfonate hydrate 98%
Sodium-3-[ethyl(m-tolyl)amino]-2-hydroxy-1-propanesulfonate hydrate

Precaución: for Research Use Only

Formula: C₁₂H₁₉NO₄S
Storage Temperature: Ambient
MDL Number: MFCD00040641
CAS Number: 82692-93-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR