3-(Methylthio)-2,4-triazine 98%

Supplier: Ambeed
Warning

A319225-1G A319225-25G A319225-5G A319225-100G A319225-10G
103825-194EA 6 USD
103825-194 103836-126 76838-976 76838-978 76838-980 77290-266 77616-406
3-(Methylthio)-2,4-triazine 98%
3-(Methylthio)-2,4-triazine

Precaución: for Research Use Only

Formula: C₄H₅N₃S
Boiling Pt: 275.7 °C (760 mm Hg)
Flash Pt: 120.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00490024
CAS Number: 28735-21-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR