3-(Methylsulfonylamino)phenylboronic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Methylsulphonylaminophenylboronic acid, 3-(Methylsulphonylamino)phenylboronic Acid
A379575-1G
A379575-250MG
103826-650EA
18
USD
103826-650
103837-686
76840-998
77003-628
3-(Methylsulfonylamino)phenylboronic acid 98%
3-(Methylsulfonylamino)phenylboronic acid
Precaución: for Research Use Only
Formula:
C₇H₁₀BNO₄S Boiling Pt: 433.7 °C (760 mm Hg) Melting Pt: 90…96 °C Flash Pt: 216.1 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD02179478 CAS Number: 148355-75-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...