N-(3-Aminophenyl)methanesulfonamide 98%

Supplier: Ambeed
Warning

A541821-10G A541821-1G A541821-5G
103828-636EA 14 USD
103828-636 103839-756 76726-370 76843-810 77307-772
N-(3-Aminophenyl)methanesulfonamide 98%
N-(3-Aminophenyl)methanesulfonamide

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O₂S
Boiling Pt: 361.8 °C (760 mm Hg)
Melting Pt: 117…121 °C
Flash Pt: 172.6 °C
Storage Temperature: Ambient
MDL Number: MFCD03094037
CAS Number: 37045-73-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR