3-(Methylamino)-1-propanol 98%

Supplier: Ambeed
Danger

Synonyms: 3-(Methylamino)propan-1-ol

A815731-1G A815731-10G
103841-106EA 17 USD
103841-106 76845-348 76981-710
3-(Methylamino)-1-propanol 98%
3-(Methylamino)-1-propanol

Precaución: For research use only

Formula: C₄H₁₁NO
MW: 89.14 g/mol
Boiling Pt: 166…170 °C (760 mm Hg)
Flash Pt: 82.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06797456
CAS Number: 42055-15-2
UN: 2735
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR