3-(Methoxycarbonyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-boronobenzoate, 3-(Methoxycarbonyl)benzeneboronic acid

A225713-1G A225713-25G A225713-5G A225713-10G A225713-100G
103819-192EA 5 USD
103819-192 103850-092 77088-796 77088-798 77088-800 77249-486 77615-876
3-(Methoxycarbonyl)phenylboronic acid 98%
3-(Methoxycarbonyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₉BO₄
MW: 179.97 g/mol
Boiling Pt: 359.9 °C (760 mm Hg)
Melting Pt: 205…208 °C
Flash Pt: 171.5 °C
Storage Temperature: Ambient
MDL Number: MFCD02093046
CAS Number: 99769-19-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR