3-(Methoxycarbonyl)bicyclo[1.1.1]pentane-1-carboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: Bicyclo[1.1.1]pentane-1,3-dicarboxylic acid 1-Monomethyl ester

A387227-1G A387227-5G A387227-100G A387227-100MG A387227-250MG A387227-25G
103826-820EA 19 USD
103826-820 103837-862 76841-222 76841-224 76990-074 76990-076 76990-078 76990-080
3-(Methoxycarbonyl)bicyclo[1.1.1]pentane-1-carboxylic acid 95%
3-(Methoxycarbonyl)bicyclo[1.1.1]pentane-1-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₁₀O₄
MW: 170.16 g/mol
Boiling Pt: 272.2±40.0 °C (760 mm Hg)
Flash Pt: 111.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD20638314
CAS Number: 83249-10-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR