Methyl hydrogen isophthalate 97%

Supplier: Ambeed
Warning

Synonyms: Isophthalic acid monomethyl ester, 3-(Methoxycarbonyl)benzoic acid, Benzene-1,3-dicarboxylic acid monomethyl ester, mono-Methyl isophthalate

A269264-1G A269264-25G A269264-5G A269264-100G A269264-10G
103812-958EA 5 USD
103812-958 103852-864 77092-622 77092-624 77092-626 77098-970 77098-972
Methyl hydrogen isophthalate 97%
Methyl hydrogen isophthalate

Precaución: for Research Use Only

Formula: C₉H₈O₄
Melting Pt: 194…196 °C
Storage Temperature: Ambient
MDL Number: MFCD00029972
CAS Number: 1877-71-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR