3-(Methoxycarbonyl)-2-nitrobenzoic acid 97%
Supplier: Ambeed
|
Warning
|
A315352-250MG
A315352-1G
A315352-5G
103825-062EA
29
USD
103825-062
103835-988
77277-606
77277-608
77277-610
77306-128
77306-130
3-(Methoxycarbonyl)-2-nitrobenzoic acid 97%
3-(Methoxycarbonyl)-2-nitrobenzoic acid
Precaución: for Research Use Only
Formula:
C₉H₇NO₆ Boiling Pt: 400.9 °C (760 mm Hg) Flash Pt: 196.2 °C Storage Temperature: Ambient |
MDL Number:
MFCD08690069 CAS Number: 861593-27-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...