3-Piperidinemethanol 98%

Supplier: Ambeed
Danger

Synonyms: 3-(Hydroxymethyl)piperidine

A774319-25G A774319-10G
103840-948EA 9 USD
103840-948 76845-202 77293-632
3-Piperidinemethanol 98%
3-Piperidinemethanol

Precaución: for Research Use Only

Formula: C₆H₁₃NO
MW: 115.18 g/mol
Boiling Pt: 240.4 °C (760 mm Hg)
Melting Pt: 58…62 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00005997
CAS Number: 4606-65-9
UN: 3263
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR