3-(Hydroxymethyl)benzoic acid 98%

Supplier: Ambeed
Warning

A217817-1G A217817-25G A217817-5G A217817-250MG
103818-660EA 13 USD
103818-660 103849-388 77088-030 77275-386 77303-464 77748-343
3-(Hydroxymethyl)benzoic acid 98%
3-(Hydroxymethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₈O₃
Boiling Pt: 358.1 °C (760 mm Hg)
Flash Pt: 184.6 °C
Storage Temperature: Ambient
MDL Number: MFCD06208242
CAS Number: 28286-79-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR