3-(Hydroxymethyl)-1-adamantanol 98%

Supplier: Ambeed
Warning

Synonyms: 3-(Hydroxymethyl)adamantan-1-ol, 1-Hydroxy-3-(hydroxymethyl)adamantane

A245422-10G A245422-250MG A245422-1G A245422-25G A245422-5G
103811-644EA 13 USD
103811-644 103851-480 77031-354 77031-356 77090-728 77090-730 77090-732
3-(Hydroxymethyl)-1-adamantanol 98%
3-(Hydroxymethyl)-1-adamantanol

Precaución: for Research Use Only

Formula: C₁₁H₁₈O₂
MW: 182.26 g/mol
Melting Pt: 158…162 °C
Storage Temperature: Ambient
MDL Number: MFCD03105096
CAS Number: 38584-37-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR