3-(Hydroxymethyl)-1-methyl-2-pyrrolidinone 96%

Supplier: Ambeed
Warning

Synonyms: 3-(Hydroxymethyl)-1-methylpyrrolidin-2-one

A113138-100MG A113138-250MG
103798-168EA 82 USD
103798-168 103824-008 76982-830 76982-832
3-(Hydroxymethyl)-1-methyl-2-pyrrolidinone 96%
3-(Hydroxymethyl)-1-methyl-2-pyrrolidinone

Precaución: for Research Use Only

Formula: C₆H₁₁NO₂
MW: 129.157 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18816870
CAS Number: 577780-05-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR