N-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-12, SB-12) 97%

Supplier: Ambeed
Warning

Synonyms: sulphobetaine-12, 3-(Dodecyldimethylammonio)propane-1-sulphonate, N-Dodecyl-n,n-dimethyl-3-ammonio-1-propanesulphonate

A154970-100G A154970-10G A154970-1G A154970-25G A154970-5G
103804-698EA 9 USD
103804-698 103842-894 76719-900 76719-902 76719-904 77318-626 77318-628 77318-630
N-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-12, SB-12) 97%
N-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-12, SB-12)

Precaución: for Research Use Only

Formula: C₁₇H₃₇NO₃S
MW: 335.55 g/mol
Boiling Pt: 250…260 °C (760 mm Hg)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00036909
CAS Number: 14933-08-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR