3-(Difluoromethyl)-1-methyl-1H-pyrazole-4-carboxylic acid 98%

Supplier: Ambeed
Warning

A104307-100G A104307-10G A104307-250MG A104307-25G A104307-500G A104307-1G A104307-5G
103796-686EA 6 USD
103796-686 103822-396 76970-324 76970-326 76970-328 76970-330 76970-332 77309-264 77309-266
3-(Difluoromethyl)-1-methyl-1H-pyrazole-4-carboxylic acid 98%
3-(Difluoromethyl)-1-methyl-1H-pyrazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₆F₂N₂O₂
MW: 176.121 g/mol
Boiling Pt: 315.9 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD11865267
CAS Number: 176969-34-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR